missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4-chlorobenzoate, 98%

Product Code. 11387566
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387566 10 g 10g
11397566 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387566 Supplier Thermo Scientific Alfa Aesar Supplier No. L11125.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5900-58-3 | C8H8ClNO2 | 185.607 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5900-58-3
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00017568
InChI Key YPSSCICDVDOEAI-UHFFFAOYSA-N
Synonym methyl 4-chloroanthranilate, 2-amino-4-chloro-benzoic acid methyl ester, methyl2-amino-4-chlorobenzoate, benzoic acid, 2-amino-4-chloro-, methyl ester, 2-amino-4-chlorobenzoic acid methyl ester, pubchem4643, methyl 4-chloro-anthranilate, methyl 2-amino4-chlorobenzoate, methyl-2-amino-4-chlorobenzoate
PubChem CID 80001
IUPAC Name methyl 2-amino-4-chlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Cl)N

Specifications

Melting Point 66°C to 68°C
Quantity 10 g
Beilstein 1072895
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4-chlorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.