missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3,5-dibromobenzoate, 98+%, Thermo Scientific™

Product Code. 11480707
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480707 100 g 100g
11460707 5 g 5g
11470707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480707 Supplier Thermo Scientific Alfa Aesar Supplier No. B22518.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-00-8
Molecular Formula C8H7Br2NO2
Molecular Weight (g/mol) 308.96
MDL Number MFCD00010873
InChI Key NGXVMFCGYYHEGC-UHFFFAOYSA-N
Synonym methyl 3,5-dibromoanthranilate, benzoic acid, 2-amino-3,5-dibromo-, methyl ester, 3,5-dibromoanthranilic acid methyl ester, 2-amino-3,5-dibromobenzoic acid methyl ester, anthranilic acid, 3,5-dibromo-, methyl ester, methyl 2-amino-3,5-dibromo-benzoate, pubchem3352, acmc-209mkw, ksc490q2l, methyl,3,5-dibromoanthranilate
PubChem CID 69069
IUPAC Name methyl 2-amino-3,5-dibromobenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1N

Specifications

Melting Point 87°C to 89°C
Quantity 100 g
Beilstein 3257505
Formula Weight 308.97
Percent Purity ≥98%
Chemical Name or Material Methyl 2-amino-3,5-dibromobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.