missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dihydroxybenzoate, 97%, Thermo Scientific™

Product Code. 11935101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11935101 5 g 5g
11955101 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935101 Supplier Thermo Scientific Acros Supplier No. 442450050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2150-45-0
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00059619
InChI Key WCQZCKUNZVMBDC-UHFFFAOYSA-N
Synonym 2,6-dihydroxybenzoic acid methyl ester, benzoic acid, 2,6-dihydroxy-, methyl ester, .gamma.-resorcylic acid, methyl ester, acmc-209fl9, 2-methoxycarbonyl resorcinol, methyl 2,6-dihydroxy benzoate, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate, 2-methoxycarbonyl benzene-1,3-diol
PubChem CID 519869
IUPAC Name methyl 2,6-dihydroxybenzoate
SMILES COC(=O)C1=C(O)C=CC=C1O

Specifications

Melting Point 67°C to 69°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 2, 6-dihydroxybenzoate
compliance-icons
Product Identifier
  • Methyl 2,6-dihydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.