missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dichloropyridine-4-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15453157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453157 5 g 5g
15443157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453157 Supplier Thermo Scientific Alfa Aesar Supplier No. A17468.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Sparingly Soluble (0.13 g/L) (25°C)

Notes
Keep away from oxidizing agents, reducing agents, bases, amines. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42521-09-5
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00044409
InChI Key XSKGHSUHOYEBTK-UHFFFAOYSA-N
PubChem CID 93237
IUPAC Name methyl 2,6-dichloropyridine-4-carboxylate
SMILES COC(=O)C1=CC(Cl)=NC(Cl)=C1

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Synonym methyl 2,6-dichloroisonicotinate, methyl2,6-dichloroisonicotinate, 2,6-chloro-4-pyridinecarboxylic acid methyl ester, 2,6-dichloroisonicotinic acid methyl ester, methyl-2,6-dichloroisonicotinate, 4-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, methyl-2,6-dichlorisonicotinat, acmc-209yt4, xskghsuhoyebtk-uhfffaoysa, methyl 2,6-dichloro-isonicotinate
Solubility Information Sparingly Soluble (0.13g/L) (25°C)
Formula Weight 206.03
Percent Purity 97%
Chemical Name or Material Methyl 2,6-dichloropyridine-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.