missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dichlorophenylacetate, 99%

Product Code. 10384442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10384442 5 g Glass bottle 5g
10058811 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10384442 Supplier Thermo Scientific Acros Supplier No. 351070050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54551-83-6 | C9H8Cl2O2 | 219.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54551-83-6
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.07
MDL Number MFCD00191640
InChI Key FCWRUYPZZJPCCG-UHFFFAOYSA-N
Synonym methyl 2,6-dichlorophenylacetate, methyl 2-2,6-dichlorophenyl acetate, 2,6-dichlorophenylacetic acid methyl ester, benzeneacetic acid,2,6-dichloro-, methyl ester, benzeneacetic acid, 2,6-dichloro-, methyl ester, acmc-209lhf, methyl2,6-dichlorophenylacetate, guanfacine methyl ester impurity, methyl 2,6-dichlorophenyl acetate, 2,6-dichloro-phenylacetic acid methyl ester
PubChem CID 2734107
IUPAC Name methyl 2-(2,6-dichlorophenyl)acetate
SMILES COC(=O)CC1=C(C=CC=C1Cl)Cl

Specifications

Color Colorless
Density 1.3300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.542
Quantity 5 g
Specific Gravity 1.33
Formula Weight 219.07
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 2, 6-dichlorophenylacetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2,6-dichlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.