missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dichloro-5-fluoronicotinate, 90%, Thermo Scientific™

Product Code. 10283462
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 189281-66-1
Molecular Formula C7H4Cl2FNO2
Molecular Weight (g/mol) 224.01
MDL Number MFCD02180634
InChI Key WADLLLSMEPLCNO-UHFFFAOYSA-N
Synonym methyl 2,6-dichloro-5-fluoronicotinate, 3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro-, methyl ester, 3-pyridinecarboxylicacid, 2,6-dichloro-5-fluoro-, methyl ester, 2,6-dichloro-5-fluoro-nicotinic acid methyl ester, 2,6-dichloro-3-fluoro-5-methoxycarbonyl pyridine, methyl 2,6-dichloro-5-fluoro-3-pyridinecarboxylate, 2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid methyl ester, methyl 2,6-bis chloranyl-5-fluoranyl-pyridine-3-carboxylate
PubChem CID 2775354
IUPAC Name methyl 2,6-dichloro-5-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CC(F)=C(Cl)N=C1Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 224.02
Percent Purity 90%
Chemical Name or Material Methyl 2, 6-dichloro-5-fluoronicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'