missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dichloro-5-fluoronicotinate, 90%, Thermo Scientific™

Product Code. 10079311
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10079311 10 g Amber Glass Bottle 10g
10283462 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10079311 Supplier Thermo Scientific Maybridge Supplier No. RH02135EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 189281-66-1
Molecular Formula C7H4Cl2FNO2
Molecular Weight (g/mol) 224.01
MDL Number MFCD02180634
InChI Key WADLLLSMEPLCNO-UHFFFAOYSA-N
Synonym methyl 2,6-dichloro-5-fluoronicotinate, 3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro-, methyl ester, 3-pyridinecarboxylicacid, 2,6-dichloro-5-fluoro-, methyl ester, 2,6-dichloro-5-fluoro-nicotinic acid methyl ester, 2,6-dichloro-3-fluoro-5-methoxycarbonyl pyridine, methyl 2,6-dichloro-5-fluoro-3-pyridinecarboxylate, 2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid methyl ester, methyl 2,6-bis chloranyl-5-fluoranyl-pyridine-3-carboxylate
PubChem CID 2775354
IUPAC Name methyl 2,6-dichloro-5-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CC(F)=C(Cl)N=C1Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 224.02
Percent Purity 90%
Chemical Name or Material Methyl 2, 6-dichloro-5-fluoronicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.