missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate, 95%, Thermo Scientific™

Product Code. 10255424
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10255424 25 g 25g
10570743 1 g 1g
10315162 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255424 Supplier Thermo Scientific Maybridge Supplier No. SP00004EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69687-80-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00203859
InChI Key OQWZEJIISPYZPW-UHFFFAOYSA-N
Synonym methyl 2,5-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-, methyl ester, 2,5-dimethyl-1h-pyrrole-3-carboxylic acid methyl ester, maybridge1_008495, acmc-1b2uc, 2,5-dimethyl,4-carbomethoxy pyrrole, 2,5-dimethylpyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylicacid,2,5-dimethyl-,methyl ester, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-, methyl ester
PubChem CID 592729
IUPAC Name methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
SMILES CC1=CC(=C(N1)C)C(=O)OC

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 25 g
Formula Weight 153.18
Percent Purity 95%
Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.