missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dichlorothiophene-3-carboxylate, 98%

Product Code. 11337995
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337995 5 g 5g
11327995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337995 Supplier Thermo Scientific Alfa Aesar Supplier No. L08118.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 145129-54-0 | C6H4Cl2O2S | 211.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145129-54-0
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.056
MDL Number MFCD00051763
InChI Key VCXPHMKCDRPIDG-UHFFFAOYSA-N
Synonym acmc-1c2sm, methyl 2,5-dichloro-3-thiophenecarboxylate, methyl-2,5-dichlorothiophene-3-carboxylate, methyl 2,5-dichloro-3-thiophenecarboxylate #, methyl 2,5-bis chloranyl thiophene-3-carboxylate, 2,5-dichloro-3-thiophenecarboxylic acid methyl ester, 3-thiophenecarboxylic acid, 2,5-dichloro-, methyl ester, 3-thiophenecarboxylicacid, 2,5-dichloro-, methyl ester
PubChem CID 518961
IUPAC Name methyl 2,5-dichlorothiophene-3-carboxylate
SMILES COC(=O)C1=C(SC(=C1)Cl)Cl

Specifications

Boiling Point 74°C to 75°C (0.6 mmHg)
Refractive Index 1.563
Quantity 5 g
Formula Weight 211.07
Percent Purity 98%
Chemical Name or Material Methyl 2,5-dichlorothiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.