missing translation for 'onlineSavingsMsg'
Learn More

methyl 2-(4-isobutylphenyl)propanoate, Thermo Scientific™

Product Code. 10346972
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346972 10 g Amber Glass Bottle 10g
10631755 1 g Amber Glass Bottle 1g
10438704 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10346972 Supplier Thermo Scientific Maybridge Supplier No. SEW06433EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

methyl 2-(4-isobutylphenyl)propanoate

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61566-34-5
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
InChI Key YNZYUHPFNYBBFF-UHFFFAOYSA-N
Synonym ibuprofen methyl ester, methyl 2-4-isobutylphenyl propanoate, motrin methyl ester, methyl 2-4-isobutylphenyl propionate, methyl 2-4-2-methylpropyl phenyl propanoate, 2-4-isobutylphenyl propionic acid methyl ester, racemic ibuprofen methyl ester, s-+-ibuprofen methyl ester, 2r/s-2-4-isobutylphenyl-propionic acid methyl ester, benzeneacetic acid,, a-methyl-4-2-methylpropyl-,methyl ester
PubChem CID 109101
IUPAC Name methyl 2-[4-(2-methylpropyl)phenyl]propanoate
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(=O)OC

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material Methyl 2-(4-isobutylphenyl)propanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.