missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dimethoxybenzoate, 98%, Thermo Scientific™

Product Code. 10535524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10535524 5 g 5g
10471084 25 g 25g
10021314 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10535524 Supplier Thermo Scientific Acros Supplier No. 426810050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2150-41-6
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00070649
InChI Key IHIJFZWLGPEYPJ-UHFFFAOYSA-N
PubChem CID 137451
IUPAC Name methyl 2,4-dimethoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C=C1OC

Specifications

Density 1.1940g/mL
Boiling Point 294°C to 296°C
Flash Point >110°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH30)2C6H3CO2CH3
Quantity 5 g
Specific Gravity 1.194
Synonym 2,4-dimethoxybenzoic acid methyl ester, benzoic acid, 2,4-dimethoxy-, methyl ester, acmc-1ccii, methyl2,4-dimethoxybenzoate, methyl 2,4-dimethoxybenzoate, 2,4-dimethoxy-benzoic acid methyl ester, benzoic acid,2,4-dimethoxy-,methyl ester, benzoic acid,2,4-dimethoxy-, methyl ester, 2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.