missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%

Product Code. 12387173 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12387173 5 g 5g
12397173 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12387173 Supplier Thermo Scientific Acros Supplier No. 446580050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6299-85-0 | C6H4Cl2N2O2 | 207.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6299-85-0
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.02
InChI Key DQNQNLWKAGZNIT-UHFFFAOYSA-N
Synonym methyl 2,4-dichloropyrimidine-6-carboxylate, methyl 2,4-dichloro-pyrimidine-6-carboxylate, 2,6-dichloropyrimidine-4-carboxylic acid methyl ester, 4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester, 2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester, t6n cnj bg dvo1 fg wln, zlchem 33, pubchem23452, methyl2,6-dichloropyrimidine-4-carboxylate, 2,4-dichloro-6-methoxycarbonyl pyrimidine
PubChem CID 239689
IUPAC Name methyl 2,6-dichloropyrimidine-4-carboxylate
SMILES COC(=O)C1=CC(=NC(=N1)Cl)Cl

Specifications

Melting Point 55°C to 56°C
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material Methyl 2, 4-dichloropyrimidine-6-carboxylate
compliance-icons
Product Identifier
  • Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.