missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dichlorobenzoate, 97%, Thermo Scientific™

Product Code. 10068369 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10068369 25 g 25g
10027989 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10068369 Supplier Thermo Scientific Acros Supplier No. 430050250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 35112-28-8
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.04
MDL Number MFCD00085117
InChI Key VCRWILYAWSRHBN-UHFFFAOYSA-N
Synonym methyl2,4-dichlorobenzoate, 2,4-dichlorobenzoic acid methyl ester, benzoic acid, 2,4-dichloro-, methyl ester, methyl-2,4-dichlorobenzoate, pubchem3713, methyl ester of 2,4-dichlorobenzoic acid, acmc-209id8, ksc497m5b, methyl 2,4-dichloro benzoate
PubChem CID 37055
IUPAC Name methyl 2,4-dichlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

Color Colorless to Yellow
Boiling Point 132.0°C (15.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 205.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 2,4-dichlorobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2,4-dichlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.