missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dichlorobenzoate, 97%, Thermo Scientific™

Product Code. p-7025209
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408987 2.5 g 2.5g
11418987 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408987 Supplier Thermo Scientific Alfa Aesar Supplier No. B20583.88

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35112-28-8
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00085117
InChI Key VCRWILYAWSRHBN-UHFFFAOYSA-N
Synonym methyl2,4-dichlorobenzoate, 2,4-dichlorobenzoic acid methyl ester, benzoic acid, 2,4-dichloro-, methyl ester, methyl-2,4-dichlorobenzoate, pubchem3713, methyl ester of 2,4-dichlorobenzoic acid, acmc-209id8, ksc497m5b, methyl 2,4-dichloro benzoate
PubChem CID 37055
IUPAC Name methyl 2,4-dichlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

Density 1.386
Boiling Point 131°C to 133°C (15 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.557
Quantity 2.5 g
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material Methyl 2,4-dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.