missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4,6-trihydroxybenzoate, 98%

Product Code. 11415053
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415053 25 g 25g
11405053 5 g 5g
11425053 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415053 Supplier Thermo Scientific Alfa Aesar Supplier No. A12305.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3147-39-5 | C8H8O5 | 184.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3147-39-5
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.147
MDL Number MFCD00013969
InChI Key AQDIJIAUYXOCGX-UHFFFAOYSA-N
PubChem CID 76600
IUPAC Name methyl 2,4,6-trihydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 175°C to 179°C
Quantity 25 g
Beilstein 2837819
Synonym benzoic acid, 2,4,6-trihydroxy-, methyl ester, 2,4,6-trihydroxybenzoic acid methyl ester, acmc-20akj4, benzoic acid,2,4,6-trihydroxy-, methyl ester
Formula Weight 184.15
Percent Purity 98%
Chemical Name or Material Methyl 2,4,6-trihydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.