missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Mikromol™

Product Code. 30461327
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461327 4 x 25 mg 100mg
30051725 100 mg 100mg
30052264 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30461327 Supplier Mikromol Supplier No. MM0505.0400254AMP
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate
CAS 42019-07-8
Analyte or Component Names Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate
Shipping Condition Room Temperature
Molecular Formula C18 H17 Cl O4
Certifications/Compliance ISO 17025
Synonym Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Fenofibrate Imp. D (EP), Fenofibrate Impurity D
InChI Formula InChI=1S/C18H17ClO4/c1-18(2,17(21)22-3)23-15-10-6-13(7-11-15)16(20)12-4-8-14(19)9-5-12/h4-11H,1-3H3
Solution Type Neat
SMILES COC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2
IUPAC Name methyl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
Molecular Weight (g/mol) 332.78
Quantity 4 x 25 mg
Formula Weight 332.0815 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.