missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3-dichloropropionate, 98%

Product Code. 11459467
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459467 25 g 25g
11469467 100 g 100g
11479467 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459467 Supplier Thermo Scientific Alfa Aesar Supplier No. B21993.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3674-09-7 | C4H6Cl2O2 | 156.99 g/mol

Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.

Solubility
Soluble in ether, acetone, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3674-09-7
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 156.99
MDL Number MFCD00000944
InChI Key OFHMODDLBXETIK-UHFFFAOYNA-N
IUPAC Name methyl 2,3-dichloropropanoate

Specifications

Density 1.326
Boiling Point 91°C to 92°C (50 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.453
Quantity 25 g
UN Number UN3265
Beilstein 1701891
Synonym methyl vinyl sulfone, ethene, methylsulfonyl, methyl vinyl sulphone, methylvinylsulfone, methanesulfonylethene, sulfone, methyl vinyl, vinyl methyl sulfone, methanesulfonyl-ethene, methylsulfonyl ethene, methyl vinyl sulfphone
Solubility Information Soluble in ether,acetone,ethanol.
SMILES COC(=O)C(Cl)CCl
PubChem CID 19354
Formula Weight 157
Percent Purity 98%
Chemical Name or Material Methyl 2,3-dichloropropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.