missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(3-chloropropanoylamino)-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15425155
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15425155

Brand: Thermo Scientific Alfa Aesar H33025.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551910-46-4
Molecular Formula C12H14ClNO3S
Molecular Weight (g/mol) 287.758
MDL Number MFCD01354347
InChI Key OMCRFVOYMSUXGQ-UHFFFAOYSA-N
Synonym methyl 2-3-chloropropanamido-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, methyl 2-3-chloropropanoylamino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, methyl 2-3-chloropropanamido-4h,5h,6h-cyclopenta b thiophene-3-carboxylate
PubChem CID 4982506
IUPAC Name methyl 2-(3-chloropropanoylamino)-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
SMILES COC(=O)C1=C(SC2=C1CCC2)NC(=O)CCCl

Specifications

Melting Point 108°C to 109°C
Quantity 5 g
Formula Weight 287.77
Percent Purity 96%
Chemical Name or Material Methyl 2-(3-chloropropanoylamino)-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.