missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%

Product Code. 10576864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10576864 100 g Glass bottle 100g
10470704 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10576864 Supplier Thermo Scientific Acros Supplier No. 210911000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14002-80-3 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14002-80-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00009707
InChI Key KJRFTNVYOAGTHK-UHFFFAOYSA-N
Synonym hydroxypivalic acid methyl ester, methyl 3-hydroxypivalate, methyl hydroxypivalate, methyl 2,2-dimethyl-3-hydroxypropionate, hydroxypivalicacidmethylester, propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester, 2,2-dimethyl-3-hydroxypropionic acid methyl ester, 3-hydroxy-2,2-dimethyl-propionic acid methyl ester, methyl 2,2-dimethyl-3-hydroxypropanoate, methyl hydroxylpivalate
PubChem CID 84152
IUPAC Name methyl 3-hydroxy-2,2-dimethylpropanoate
SMILES COC(=O)C(C)(C)CO

Specifications

Color Colorless
Density 1.0400g/mL
Boiling Point 177°C to 178°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2C(CH3)2CO2CH3
Quantity 100 g
Beilstein 03, 331
Specific Gravity 1.04
Formula Weight 132.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl 2,2-dimethyl-3-hydroxypropionate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2,2-dimethyl-3-hydroxypropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.