missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%

Product Code. 15470086
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470086 25 g 25g
15419487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470086 Supplier Thermo Scientific Alfa Aesar Supplier No. H52745.14
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 680-15-9 | C3H3F3O4S | 192.108 g/mol

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680-15-9
Molecular Formula C3H3F3O4S
Molecular Weight (g/mol) 192.108
MDL Number MFCD00144316
InChI Key GQJCAQADCPTHKN-UHFFFAOYSA-N
Synonym methyl 2,2-difluoro-2-fluorosulfonyl acetate, methylfluorosulphonyldifluoroacetate, methyl difluoro fluorosulfonyl acetate, methyl fluorosulfonyldifluoroacetate, methyl fluorosulphonyl difluoro acetate, methyl fluorosulphonyldifluoroacetate, methyl 2-fluorosulfonyl difluoroacetate, fluorosulfonyldifluoroacetic acid methyl ester, 2,2-difluoro-2-fluorosulfonylacetic acid methyl ester, difluoro fluorosulfonyl acetic acid methyl ester
PubChem CID 547667
IUPAC Name methyl 2,2-difluoro-2-fluorosulfonylacetate
SMILES COC(=O)C(F)(F)S(=O)(=O)F

Specifications

Density 1.509
Boiling Point 117°C to 118°C
Flash Point 47°C (117°F)
Linear Formula FSO2CF2CO2CH3
Refractive Index 1.351
Quantity 25 g
UN Number UN2920
Beilstein 1812896
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.