missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2,2-trichloroacetimidate, 98%

Product Code. 10721155 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721155 100 g Glass bottle 100g
10408360 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10721155 Supplier Thermo Scientific Acros Supplier No. 160931000
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2533-69-9 | C3H4Cl3NO | 176.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2533-69-9
Molecular Formula C3H4Cl3NO
Molecular Weight (g/mol) 176.42
MDL Number MFCD00001759
InChI Key OGBINJLTBZWRRB-UHFFFAOYSA-N
Synonym methyl 2,2,2-trichloroacetimidate, methyl trichloroacetimidate, methyl 2,2,2-trichloroethanecarboximidate, ethanimidic acid, 2,2,2-trichloro-, methyl ester, acmc-20aebo, methyltrichloroacetimidate, methyl trichloroacetoimidate, methyl-2,2,2-trichloroacetimidate, methyl 2,2,2-trichloroethanimidoate, trichloroacetimidic acid methyl ester
PubChem CID 100479
IUPAC Name methyl 2,2,2-trichloroethanimidate
SMILES COC(=N)C(Cl)(Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.4200g/mL
Boiling Point 148.0°C to 149.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CCl3C(=NH)OCH3
Refractive Index 1.4770 to 1.4790
Quantity 100 g
Beilstein 02, 212
Specific Gravity 1.42
Formula Weight 176.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2,2,2-trichloroacetimidate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2,2,2-trichloroacetimidate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.