missing translation for 'onlineSavingsMsg'
Learn More

Methyl-1-naphthalenemethylamine-D3, TRC

Product Code. 30467484
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467484 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467484 Supplier Toronto Research Chemicals Supplier No. TRCM3205625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl-1-naphthalenemethylamine-d3
CAS 1033720-13-6
Molecular Formula C12H10D3N
Quantity 5 mg
Synonym N-Methyl-1-naphthalenemethanamine, [(Naphthalen-1-yl)methyl]methylamine, N-Methyl-N-[(naphthalen-1-yl)methyl]amine, N-Methyl-N-[(1-naphthyl)methyl]amine, 1-Methylaminomethylnaphthalene, NSC 129392
InChI Formula InChI=1S/C12H13N/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11/h2-8,13H,9H2,1H3/i1D3
SMILES [2H]C([2H])([2H])NCc1cccc2ccccc12
IUPAC Name 1,1,1-trideuterio-N-(naphthalen-1-ylmethyl)methanamine
Molecular Weight (g/mol) 174.26
Formula Weight 174.1236
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.