missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylcyclopropyl ketone, 96%

Product Code. 15428839
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15428839 Supplier Thermo Scientific Alfa Aesar Supplier No. L05394.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1567-75-5 | C6H10O | 98.145 g/mol

Methyl 1-methylcyclopropyl ketone is used as a pharmaceutical intermediate and an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-methylcyclopropyl ketone is used as a pharmaceutical intermediate and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly
TRUSTED_SUSTAINABILITY

Specifications

CAS 1567-75-5
Density 0.896
Boiling Point 126°C to 128°C
Flash Point 23°C (73°F)
Molecular Formula C6H10O
Refractive Index 1.434
MDL Number MFCD00010245
Quantity 5 g
UN Number UN1224
Beilstein 2038600
Synonym methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone
Solubility Information Insoluble in water.
InChI Key OQBCJXUAQQMTRW-UHFFFAOYSA-N
SMILES CC(=O)C1(CC1)C
IUPAC Name 1-(1-methylcyclopropyl)ethanone
Molecular Weight (g/mol) 98.145
PubChem CID 74067
Formula Weight 98.15
Percent Purity 96%
Chemical Name or Material Methyl 1-methylcyclopropyl ketone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.