missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylcyclopropyl ketone, 95%

Product Code. 10307030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307030 25 g Glass bottle 25g
10103580 5 g Glass bottle 5g
11904841 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307030 Supplier Thermo Scientific Acros Supplier No. 147630250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1567-75-5 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1567-75-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
InChI Key OQBCJXUAQQMTRW-UHFFFAOYSA-N
Synonym methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone
PubChem CID 74067
IUPAC Name 1-(1-methylcyclopropyl)ethanone
SMILES CC(=O)C1(CC1)C

Specifications

Color Colorless to Yellow
Density 0.8950g/mL
Boiling Point 125.0°C to 128.0°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C3H4COCH3
Refractive Index 1.4330 to 1.4350
Quantity 25 g
Specific Gravity 0.895
Solubility Information Solubility in water: insoluble.
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 1-methylcyclopropyl ketone
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 1-methylcyclopropyl ketone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.