missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-cyclopentene-1-carboxylate, 95%

Product Code. 10224954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10224954 10 g Glass bottle 10g
10214954 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10224954 Supplier Thermo Scientific Acros Supplier No. 397590100
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25662-28-6 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25662-28-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00239506
InChI Key VTYCAXIAUKEGBQ-UHFFFAOYSA-N
Synonym methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487
PubChem CID 549129
IUPAC Name methyl cyclopentene-1-carboxylate
SMILES COC(=O)C1=CCCC1

Specifications

Color White to Yellow
Density 1.0310g/mL
Boiling Point 76°C to 78°C (9.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H7COOCH3
Quantity 10 g
Specific Gravity 1.031
Formula Weight 126.15
Percent Purity 97%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material Methyl 1-cyclopentene-1-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 1-cyclopentene-1-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.