missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-cyclohexene-1-carboxylate, 98%

Product Code. 10476232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10476232 5 g Glass bottle 5g
10773773 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10476232 Supplier Thermo Scientific Acros Supplier No. 213710050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18448-47-0 | C8H12O2 | 140.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18448-47-0
MDL Number MFCD00001544
InChI Key KXPWRCPEMHIZGU-UHFFFAOYSA-N
Synonym methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene
PubChem CID 87647
IUPAC Name methyl cyclohexene-1-carboxylate
SMILES COC(=O)C1=CCCCC1

Specifications

Color Colorless
Density 1.0300g/mL
Boiling Point 190°C to 192°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H12O2
Linear Formula C6H9CO2CH3
Refractive Index 1.4755 to 1.4775
Quantity 5 g
Beilstein 09, IV, 112
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Molecular Weight (g/mol) 140.18
Formula Weight 140.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 1-cyclohexene-1-carboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.