missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 98%, Thermo Scientific™

Product Code. 10614872 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10614872

Brand: Thermo Scientific Acros 179740050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 51535-00-3
Melting Point 62°C to 66°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C13H15NO3
MDL Number MFCD00003196
Quantity 5 g
Synonym methyl 1-benzyl-5-oxo-3-pyrrolidinecarboxylate, 1-benzyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester, methyl 1-benzyl-5-oxo-pyrrolidine-3-carboxylate, methyl1-benzyl-5-oxo-3-pyrrolidinecarboxylate, 3-pyrrolidinecarboxylic acid, 5-oxo-1-phenylmethyl-, methyl ester, methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 1-benzyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester, methyl 5-oxo-1-benzylpyrrolidine-3-carboxylate, pubchem17184, methyl-1-benzyl-5-oxopyrrolidin-3-carboxylat
InChI Key WTRWSSDZHQOPJI-UHFFFAOYSA-N
SMILES COC(=O)C1CC(=O)N(C1)CC2=CC=CC=C2
IUPAC Name methyl 1-benzyl-5-oxopyrrolidine-3-carboxylate
Molecular Weight (g/mol) 233.27
PubChem CID 98909
Formula Weight 233.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.