missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, Thermo Scientific™

Product Code. 10804151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10804151 100 mg 100mg
10814151 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10804151 Supplier Thermo Scientific Acros Supplier No. 436621000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 72784-42-0
Molecular Formula C5H9NO2·ClH
Molecular Weight (g/mol) 151.59
InChI Key CHSDCWKLSHPKFY-UHFFFAOYSA-N
Synonym methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, methyl 1-aminocyclopropane-1-carboxylate hcl, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropanecarboxylate hcl, methyl-1-aminocyclopropanecarboxylate hydrochloride
PubChem CID 12760695
IUPAC Name methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
SMILES COC(=O)C1(CC1)N.Cl

Specifications

Melting Point 176.4°C to 178.7°C
Color White
Elemental Analysis 38.37 to 40.33% C, 8.95 to 9.40%
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 100 mg
Formula Weight 151.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-aminocyclopropanecarboxylate hydrochloride
compliance-icons
Product Identifier
  • Methyl 1-aminocyclopropanecarboxylate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.