missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,4-Benzodioxan-2-carboxylate, TRC

Product Code. 17904375
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904375 250 mg 250mg
17933305 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17904375 Supplier Toronto Research Chemicals Supplier No. TRCM289780250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3663-79-4
Molecular Formula C10 H10 O4
Molecular Weight (g/mol) 194.18
Synonym 1,4-Benzodioxin-2-carboxylic acid, 2,3-dihydro-, methyl ester, 1,4-Benzodioxan-2-carboxylic acid, methyl ester (6CI,7CI,8CI), Methyl 1,4-benzodioxan-2-carboxylate, Methyl 1,4-benzodioxane-2-carboxylate, Methyl (2RS)-1,4-benzodioxane-2-carboxylate
IUPAC Name methyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate
SMILES COC(=O)C1COc2ccccc2O1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H10O4/c1-12-10(11)9-6-13-7-4-2-3-5-8(7)14-9/h2-5,9H,6H2,1H3
Formula Weight 194.0579
Chemical Name or Material Methyl (2RS)-1,4-Benzodioxane-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.