missing translation for 'onlineSavingsMsg'
Learn More

Methoxytrimethylsilane, 97+%

Product Code. 11388978
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388978 50 g 50g
11308988 10 g 10g
11398978 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388978 Supplier Thermo Scientific Alfa Aesar Supplier No. 042464.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1825-61-2 | C4H12OSi | 104.22 g/mol

Methoxytrimethylsilane plays an important role in the synthesis of organic compounds and used as water disgusting. It is used in the preparation of alfa-lithioalkoxysilanes by treating with tert-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1825-61-2
Molecular Formula C4H12OSi
Molecular Weight (g/mol) 104.22
MDL Number MFCD00008344,MFCD03458977
InChI Key POPACFLNWGUDSR-UHFFFAOYSA-N
Synonym trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn
PubChem CID 15771
IUPAC Name methoxy(trimethyl)silane
SMILES CO[Si](C)(C)C

Specifications

Boiling Point 57°C to 58°C
Physical Form Liquid
Quantity 50 g
UN Number UN1993
Flash Point −11°C (−12°F)
Solubility Information Miscible with cyclopentane.
Formula Weight 104.22
Percent Purity ≥97%
Odor Mild
Refractive Index 1.3678
Sensitivity Moisture sensitive
Density 0.756 g/mL
Chemical Name or Material Methoxytrimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.