missing translation for 'onlineSavingsMsg'
Learn More

Methoxytributyltin, TRC

Product Code. 30469494
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30469494 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30469494

Brand: Toronto Research Chemicals TRCM51298010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methoxytributyltin
CAS 1067-52-3
Molecular Formula C13H30OSn
Quantity 10 g
Synonym TributylmethoxytinTributylmethoxy StannaneNSC 202857Tributyltin Methoxide
InChI Formula InChI=1S/3C4H9.CH3O.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;1H3;/q;;;-1;+1
SMILES CCCC[Sn](CCCC)(CCCC)OC
IUPAC Name tributyl(methoxy)stannane
Molecular Weight (g/mol) 321.09
Formula Weight 322.1319
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.