missing translation for 'onlineSavingsMsg'
Learn More

(±)-α-Methoxyphenylacetic acid, Honeywell Fluka™

Product Code. 15626130 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15626130

Brand: Honeywell-Fluka 6521025G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for gravimetric determination of Na,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7021-09-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00064217
InChI Key DIWVBIXQCNRCFE-UHFFFAOYNA-N
PubChem CID 107202
IUPAC Name 2-methoxy-2-phenylacetic acid

Specifications

Packaging Plastic Bottle
Linear Formula C6H5CH(OCH3)COOH
Quantity 25 g
UN Number NONH for all modes of transport
Beilstein 2209197
Synonym methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O
Formula Weight 166.17g/mol
Percent Purity ≥99% (T)
Chemical Name or Material (±)-α-Methoxyphenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.