missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyphenamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methoxymethamphetamine Hydrochloride |
| CAS | 5588-10-3 |
| Molecular Formula | C11 H17 N O . Cl H |
| Quantity | 50 mg |
| Synonym | Benzeneethanamine','2-methoxy-N,α-dimethyl-','hydrochloride (1:1), Benzeneethanamine','2-methoxy-N,α-dimethyl-','hydrochloride (9CI), Phenethylamine','o-methoxy-N,α-dimethyl-','hydrochloride (7CI,8CI), (±)-Methoxyphenamine hydrochloride, 2-Methoxymethamphetamine hydrochloride, Methoxyphenamine hydrochloride, Methoxyphenaminium chloride, NSC 65644, Orthoxine hydrochloride, Ortodrinex hydrochloride, Proasma, dl-Orthoxine hydrochloride, dl-o-Methoxy-N,α-dimethylphenethylamine hydrochloride, o-Methoxy-N,α-dimethylphenethylamine hydrochloride, o-Methoxymethamphetamine hydrochloride |
| InChI Formula | InChI=1S/C11H17NO.ClH/c1-9(12-2)8-10-6-4-5-7-11(10)13-3;/h4-7,9,12H,8H2,1-3H3;1H |
| SMILES | Cl.CNC(C)Cc1ccccc1OC |
| IUPAC Name | 1-(2-methoxyphenyl)-N-methylpropan-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 215.72 |
| Formula Weight | 215.1077 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?