missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetyl Chloride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Methoxyacetyl Chloride |
| CAS | 38870-89-2 |
| Molecular Formula | C3H5ClO2 |
| Quantity | 1 g |
| Synonym | 2-Methoxyacetyl Chloride, Methoxyacetic Acid Chloride, NSC 82321, α-Methoxyacetyl Chloride |
| InChI Formula | InChI=1S/C3H5ClO2/c1-6-2-3(4)5/h2H2,1H3 |
| SMILES | COCC(=O)Cl |
| Molecular Weight (g/mol) | 108.52 |
| Formula Weight | 107.9978 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction