missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetyl chloride, 97%, stab. with ca 0.3% magnesium oxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.
Solubility
Miscible with acetone, ether, carbon tetrachloride and chloroform.
Notes
Moisture sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.
Solubility
Miscible with acetone, ether, carbon tetrachloride and chloroform.
Notes
Moisture sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
Specifications
Melting Point | <-40°C |
Density | 1.185 |
Boiling Point | 112°C to 113°C |
Flash Point | 28°C (82°F) |
Refractive Index | 1.424 |
Quantity | 25 g |
UN Number | UN2920 |
Beilstein | 1740244 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with acetone,ether,carbon tetrachloride and chloroform. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'