missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetonitrile, 98%

Product Code. 10686470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10686470 100 g Glass bottle 100g
10431610 5 g Glass bottle 5g
10332500 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686470 Supplier Thermo Scientific Acros Supplier No. 125661000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1738-36-9 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1738-36-9
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00001892
InChI Key QKPVEISEHYYHRH-UHFFFAOYSA-N
PubChem CID 74442
IUPAC Name 2-methoxyacetonitrile
SMILES COCC#N

Specifications

Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 118.0°C to 119.0°C (731.0 mmHg)
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CN
Refractive Index 1.3800 to 1.3830 (20°C, 589nm)
Quantity 100 g
Beilstein 03,242
Fieser 05,436
Specific Gravity 0.95
Synonym methoxyacetonitrile, acetonitrile, methoxy, acetonitrile, 2-methoxy, ncch2och3, 2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile,2-methoxy, intermediates-zcf02294, acmc-209e7f
Formula Weight 71.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methoxyacetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methoxyacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.