missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetone, 96%

Product Code. p-3736838 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
Product Code. Quantity Packaging unitSize
10711071 10 g Glass bottle 10g
10495745 50 g Glass bottle 50g
2 options
This item is not returnable. View return policy

Product Code. 10711071

Brand: Thermo Scientific Acros 175730100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5878-19-3 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5878-19-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00008768
InChI Key CUZLJOLBIRPEFB-UHFFFAOYSA-N
Synonym methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3
PubChem CID 22172
IUPAC Name 1-methoxypropan-2-one
SMILES COCC(C)=O

Specifications

Color Yellow
Density 0.9500g/mL
Boiling Point 118.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2COCH3
Refractive Index 1.3970 to 1.3990
Quantity 10 g
Beilstein 01, 822
Specific Gravity 0.95
Solubility Information Solubility in water: miscible
Formula Weight 88.11
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methoxyacetone
Show More Show Less
compliance-icons
Product Identifier
  • Methoxyacetone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.