missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic anhydride, 95%

Product Code. 15461809
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461809 100 g 100g
15451809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461809 Supplier Thermo Scientific Alfa Aesar Supplier No. H64541.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19500-95-9 | C6H10O5 | 162.14 g/mol

Methoxyacetic anhydride is used as the intermediate of 5-Aza-2'-deoxycytidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic anhydride is used as the intermediate of 5-Aza-2′-deoxycytidine.

Solubility
Very Soluble in water.(193 g/L) (25°C)

Notes
Moisture sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19500-95-9
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD11976032
InChI Key PEHFQQWAINXOQG-UHFFFAOYSA-N
Synonym methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine
PubChem CID 88095
IUPAC Name (2-methoxyacetyl) 2-methoxyacetate
SMILES COCC(=O)OC(=O)COC

Specifications

Color Colorless
Linear Formula (CH3OCH2CO)2O
Quantity 100 g
UN Number UN3265
Beilstein 1771552
Sensitivity Moisture sensitive
Solubility Information Very Soluble in water.(193g/L) (25°C)
Formula Weight 162.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methoxyacetic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.