missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic Acid, TRC

Product Code. 30470143
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470143 1 g 1g
30474718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30470143 Supplier Toronto Research Chemicals Supplier No. TRCM2255751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-45-6
Molecular Formula C3 H6 O3
Molecular Weight (g/mol) 90.08
Synonym Acetic acid, 2-methoxy-, Acetic acid, methoxy- (6CI,7CI,8CI,9CI), 2-Methoxyacetic acid, Methoxyacetic acid, Methoxyethanoic acid, NSC 7300
IUPAC Name 2-methoxyacetic acid
SMILES COCC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H6O3/c1-6-2-3(4)5/h2H2,1H3,(H,4,5)
Formula Weight 90.0317
Chemical Name or Material Methoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.