missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic acid, 97+%

Product Code. 10781461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10781461 25 mL Glass bottle 25mL
10781651 250 mL Glass bottle 250mL
10072310 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10781461 Supplier Thermo Scientific Acros Supplier No. 186770250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-45-6 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-45-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004308
InChI Key RMIODHQZRUFFFF-UHFFFAOYSA-N
Synonym methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
PubChem CID 12251
IUPAC Name 2-methoxyacetic acid
SMILES COCC(=O)O

Specifications

Melting Point 7°C
Color Colorless to Yellow
Density 1.1740g/mL
Boiling Point 202°C to 204°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula CH3OCH2CO2H
Refractive Index 1.4153 to 1.4173
Quantity 25 mL
Beilstein 03, 232
Specific Gravity 1.174
Solubility Information Solubility in water: freely soluble
Formula Weight 90.08
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Methoxyacetic acid, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methoxyacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.