missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methotrexate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Methotrexate (Amethopterin), an antimetabolite and antifolate agent, inhibits the enzyme dihydrofolate reductase, thereby preventing the conversion of folic acid into tetrahydrofolate, and inhibiting DNA synthesis. Methotrexate, also an immunosuppressant and antineoplastic agent, is used for the research of rheumatoid arthritis and a number of different cancers (such as acute lymphoblastic leukemia).
Specifications
Specifications
| Chemical Name or Material | Methotrexate |
| CAS | 59-05-2 |
| Color | Light Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C20H22N8O5 |
| Quantity | 100 mg |
| Synonym | Amethopterin CL14377 WR19039 |
| Solubility Information | DMSO : ≥ 50 mg/mL (110.03 mM) |
| SMILES | NC1=NC(N)=C2C(N=CC(CN(C)C3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=N1 |
| Molecular Weight (g/mol) | 454.44 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction