missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methotrexate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Methotrexate |
| CAS | 59-05-2 |
| Analyte or Component Names | Methotrexate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 N8 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic acid, L-(+)-N-[p-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-glutamic acid, (+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylfolic acid, 4-Amino-N10-methylpteroylglutamic acid, Amethopterin, Amethopterine, Antifolan, CL 14377, EMT 25299, Emtexate, L-Amethopterin, L-Methotrexate, Ledertrexate, MTX, Metatrexan, Methotrexat-Ebewe, Methotrexate, Methoxtrexate, Methylaminopterin, Mexate, N-[p-[[2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-(+)-glutamic acid, NSC 740, R 9985, Rheumatrex, Trexall |
| InChI Formula | InChI=1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m0/s1 |
| Solution Type | Neat |
| SMILES | CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction