missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate, Fisher BioReagents

Product Code. 10705304
Change view
Click to view available options
Quantity:
10 mg
100 mg
Packaging:
Amber Glass
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10705304 10 mg Amber Glass 10mg
10204923 100 mg Amber Glass 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10705304 Supplier Fisher BioReagents Supplier No. BP266510
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Inhibitor of DNA and RNA synthesis
  • Inhibits the enzyme responsible for synthesizing the folic acid derivative that is necessary for purine and pyrimidine synthesis
  • The lack of purines and pyrimidines  impacts the synthesis of DNA, RNA, and proteins
  • Referenced for use in antitumor studies and other studies of biological pathways
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Decomposition Information >195°C
Melting Point 195°C
Color Yellow
Ignition Residue 0.1% max.
Assay Percent Range 98.0 to 102.0 %
Packaging Amber Glass
Quantity 10 mg
Merck Index 15, 6057
Specific Rotation +19.0 to +24.0° (+ or -)
Formula Weight 454.44
Percent Purity 98.0 to 102.0%
Physical Form Powder or Solid
Chemical Name or Material Methotrexate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methotrexate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H340-May cause genetic defects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.