missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate Antibody, Novus Biologicals™

Product Code. 18233775 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
Unit Size:
0.05mL
0.25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18233775 0.05 mL 0.05mL
18701164 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18233775 Supplier Novus Biologicals Supplier No. NB100659970.05ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Methotrexate Polyclonal specifically detects Methotrexate in Non-species specific samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methotrexate
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation Whole antisera with 0.09% Sodium Azide
Gene Alias MTX
Host Species Sheep
Immunogen Methotrexate (4-amino-10-methylpteroyl-L-glutamic acid) conjugated via the carboxyl groups of the glutamic acid residue to KLH.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65997 is specific for Methotrexate and exhibits the following cross reactivities with related compounds: Aminopterin (18%), Folic acid (<0.01%), Dyhydrofolic acid (<0.01%), Tetrahydrofolic acid (<0.01%), N-5-Methyltetrahydrofolic acid (<0.01%), Folinic acid (<0.01%), N-(p-Aminobenzoyl)-L-glutamic acid (<0.01%).
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.