missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate a-Methyl Ester (>90%), TRC

Product Code. 30132632
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132632 100 mg 100mg
30005409 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132632 Supplier Toronto Research Chemicals Supplier No. TRCM260700100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66147-29-3
Molecular Formula C21 H24 N8 O5
Molecular Weight (g/mol) 468.47
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, 1-methyl ester, Methotrexate α-methyl ester, Methotrexate 1-Methyl Ester, Methotrexate Imp. I (EP), N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic Acid 1-Methyl Ester, EP Methotrexate Impurity I
IUPAC Name (4S)-4-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]-5-methoxy-5-oxopentanoic acid
SMILES NC=1C2=C(N=CC(CN(C)C3=CC=C(C(N[C@@H](CCC(O)=O)C(OC)=O)=O)C=C3)=N2)N=C(N)N1

Specifications

InChI Formula InChI=1S/C21H24N8O5/c1-29(10-12-9-24-18-16(25-12)17(22)27-21(23)28-18)13-5-3-11(4-6-13)19(32)26-14(20(33)34-2)7-8-15(30)31/h3-6,9,14H,7-8,10H2,1-2H3,(H,26,32)(H,30,31)(H4,22,23,24,27,28)/t14-/m0/s1
Quantity 100 mg
Formula Weight 468.187
Shipping Condition Room Temperature
Chemical Name or Material Methotrexate alpha-Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.