missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal

Product Code. 30462807
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30462807

Brand: LoGiCal LGCAMP0040.0029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Methotrexate 1.0 mg/ml in Dimethyl Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methotrexate
CAS 59-05-2
Analyte or Component Names Methotrexate
Shipping Condition Ice Pack (-20°C)
Molecular Formula C20 H22 N8 O5
Certifications/Compliance ISO 17034
Synonym N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic acid, L-(+)-N-[p-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-glutamic acid, (+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylfolic acid, 4-Amino-N10-methylpteroylglutamic acid, Amethopterin, Amethopterine, Antifolan, CL 14377, EMT 25299, Emtexate, L-Amethopterin, L-Methotrexate, Ledertrexate, MTX, Metatrexan, Methotrexat-Ebewe, Methotrexate, Methoxtrexate, Methylaminopterin, Mexate, N-[p-[[2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-(+)-glutamic acid, NSC 740, R 9985, Rheumatrex, Trexall
InChI Formula InChI=1 S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13 H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m0/s1
SMILES CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2 S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Forensics
Molecular Weight (g/mol) 454.44
Quantity 1 mL
Formula Weight 454.1713 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.