missing translation for 'onlineSavingsMsg'
Learn More

Methomyl Oxime, TRC

Product Code. 30468954
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468954 25 mg 25mg
30468674 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30468954 Supplier Toronto Research Chemicals Supplier No. TRCM26054025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13749-94-5
Molecular Formula C3 H7 N O S
Molecular Weight (g/mol) 105.16
Synonym Methomyl-oxime, Acetohydroximic acid, thio-, methyl ester (8CI), 1-(Methylthio)acetaldehyde oxime, 1-(Methylthio)acetaldoxime, Methomyl oxime, Methyl N-hydroxyacetimidothioate, Methyl N-hydroxyethanimidothioate, Methyl thioacetohydroxamate, S-Methyl N-hydroxyethanimidothioate, S-Methyl N-hydroxythioacetimidate, Ethanimidothioic acid, N-hydroxy-, methyl ester
IUPAC Name methyl (1Z)-N-hydroxyethanimidothioate
SMILES CS\C(=N/O)\C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C3H7NOS/c1-3(4-5)6-2/h5H,1-2H3/b4-3-
Formula Weight 105.0248
Chemical Name or Material Methomyl-oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.