missing translation for 'onlineSavingsMsg'
Learn More

Methohexital 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30457460
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
missing translation for 'unitSize':
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30457460 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30457460 missing translation for 'mfr' LoGiCal missing translation for 'supplierNo' LGCAMP1347.0001
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methohexital
CAS 151-83-7
Analyte or Component Names Methohexital
Shipping Condition Room Temperature
Molecular Formula C14 H18 N2 O3
Certifications/Compliance ISO 17034
Synonym Methohexital
InChI Formula InChI=1S/C14H18N2O3/c1-5-7-8-10(3)14(9-6-2)11(17)15-13(19)16(4)12(14)18/h6,10H,2,5,9H2,1,3-4H3,(H,15,17,19)
Solution Type Single Solution
SMILES CCC#CC(C)C1(CC=C)C(=O)NC(=O)N(C)C1=O
IUPAC Name 5-hex-3-yn-2-yl-1-methyl-5-prop-2-enyl-1,3-diazinane-2,4,6-trione
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 262.3
Quantity 1 mL
Formula Weight 262.1317 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.