missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Method DM 471 Standard Mixture 890 100 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 88-06-2, 120-83-2, 95-57-8, 95-48-7, 108-39-4, 106-48-9 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction