Learn More
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 2-Butanone | 78-93-3 | 200μg/mL |
| 2-Chloroethyl vinyl ether | 110-75-8 | 200μg/mL |
| 2-Hexanone | 591-78-6 | 200μg/mL |
| 4-Methyl-2-pentanone | 108-10-1 | 200μg/mL |
| Acetone | 67-64-1 | 200μg/mL |
| Carbon disulfide | 75-15-0 | 200μg/mL |
| p-Xylene | 106-42-3 | 200μg/mL |
| Styrene | 100-42-5 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Volatile Organics Additional Analytes Mix D |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA Method 8240B |
| Analyte or Component Names | Acetone; 2-Butanone; Carbon disulfide; 2-Chloroethyl vinyl ether; 2-Hexanone; 4-Methyl-2-pentanone; Styrene; p-Xylene |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Element | Acetone, 2-Butanone, Carbon Disulfide, 2-Chloroethyl Vinyl Ether, 2-Hexanone, 4-Methyl-2-pentanone, Styrene, p-Xylene |
| Solvent or Matrix | Methanol-P&T |
| Quantity | 1 mL |
Sicurezza e movimentazione
- VOA Additional Analytes Mix D (High Level)
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: styrene, carbon disulphide, 4-methylpentan-2-one, hexan-2-one, methanol
Product intended for laboratory use only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.